Assay Details
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK056911.1 | - | 7 - 8 | 2059 | 152 | |
BC032837.1 | - | 7 - 8 | 2142 | 4829661 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200214400.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200214400.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200214400.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200214400.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200214400.hg.1 | TC0200015601.hg.1 | Clariom D TC |
