Assay Details
Gene Symbol: KANSL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152519.3 | NP_689732.2 | 7 - 8 | 2172 | 156 | |
XM_005246328.4 | XP_005246385.1 | 7 - 8 | 2454 | 156 | ||
XM_005246329.4 | XP_005246386.1 | 7 - 8 | 2160 | 156 | ||
XM_005246330.4 | XP_005246387.1 | 7 - 8 | 2041 | 156 | ||
XM_011510706.2 | XP_011509008.1 | 7 - 8 | 2079 | 156 | ||
XM_011510707.2 | XP_011509009.1 | 7 - 8 | 2262 | 156 | ||
XM_011510710.2 | XP_011509012.1 | 6 - 7 | 911 | 156 | ||
XM_017003430.1 | XP_016858919.1 | 7 - 8 | 2198 | 156 | ||
XM_017003431.1 | XP_016858920.1 | 3 - 4 | 4280 | 156 | ||
GenBank mRNA | AK074441.1 | - | 6 - 7 | 942 | 156 | |
AL133053.1 | - | 1 - 2 | 164 | 156 | ||
BC090854.1 | - | 6 - 7 | 1510 | 156 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002730.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
