Assay Details
| Gene Symbol: KANSL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152519.3 | NP_689732.2 | 9 - 10 | 2409 | 106 | |
| XM_005246328.4 | XP_005246385.1 | 9 - 10 | 2691 | 106 | ||
| XM_005246329.4 | XP_005246386.1 | 9 - 10 | 2397 | 106 | ||
| XM_005246330.4 | XP_005246387.1 | 9 - 10 | 2278 | 106 | ||
| XM_011510706.2 | XP_011509008.1 | 9 - 10 | 2316 | 106 | ||
| XM_011510707.2 | XP_011509009.1 | 9 - 10 | 2499 | 106 | ||
| XM_011510710.2 | XP_011509012.1 | 8 - 9 | 1148 | 106 | ||
| XM_017003430.1 | XP_016858919.1 | 9 - 10 | 2435 | 106 | ||
| XM_017003431.1 | XP_016858920.1 | 5 - 6 | 4517 | 106 | ||
| GenBank mRNA | AK074441.1 | - | 8 - 9 | 1179 | 106 | |
| AL133053.1 | - | 3 - 4 | 401 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002730.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000281772.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000415553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000418791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000428655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000429908.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000438563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000452086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000453904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000457374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | ENST00000460256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_001307976.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015601.hg.1 | NM_152519.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | ENST00000452057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | NR_110291.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010646.hg.1 | NR_110291.1 | Refseq |

KANSL1L
Genomic Map