Assay Details
Gene Symbol: RAD21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136566.2 | NP_001130038.2 | 2 - 3 | 243 | 104 | |
XM_006723602.2 | XP_006723665.1 | 2 - 3 | 280 | 104 | ||
XM_006723603.2 | XP_006723666.1 | 2 - 3 | 446 | 104 | ||
XM_006723604.2 | XP_006723667.1 | 2 - 3 | 280 | 104 | ||
XM_006723605.3 | XP_006723668.1 | 2 - 3 | 280 | 104 | ||
XM_011529317.2 | XP_011527619.1 | 2 - 3 | 442 | 104 | ||
GenBank mRNA | BC157891.1 | - | 2 - 3 | 243 | 8862712 | 104 |
BC171911.1 | - | 2 - 3 | 243 | 9054565 | 104 | |
HQ603827.1 | - | 1 - 2 | 158 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000015.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20000169.hg.1 | ENST00000381882.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000169.hg.1 | ENST00000409241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000169.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000169.hg.1 | TC20000015.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000171.hg.1 | ENST00000381882.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000171.hg.1 | ENST00000402452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000171.hg.1 | ENST00000409241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000171.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000171.hg.1 | TC20000015.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000172.hg.1 | ENST00000381882.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000172.hg.1 | ENST00000402452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000172.hg.1 | ENST00000409241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000172.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000172.hg.1 | TC20000015.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000477283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000477283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | ENST00000477283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000173.hg.1 | ENST00000381882.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000173.hg.1 | ENST00000402452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000173.hg.1 | ENST00000409241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000173.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000173.hg.1 | TC20000015.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | TC2000009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169098.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000477283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000477283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | ENST00000477283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009877.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | TC2000009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169095.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | TC2000009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169099.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | TC2000009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | ENST00000246108.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169097.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169100.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169100.hg.1 | NM_001136566.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000169100.hg.1 | TC2000009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000169100.hg.1 | ENST00000409241.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000169100.hg.1 | NM_001136566.2 | Refseq |
