Assay Details
Gene Symbol: PUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031307.3 | NP_112597.3 | 1 - 2 | 55 | 102 | |
GenBank mRNA | BM786445.1 | - | 1 - 2 | 55 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002424.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_031307.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11029061.hg.1 | ENST00000227474.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11029061.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11029061.hg.1 | TC11002424.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11029060.hg.1 | ENST00000227474.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11029060.hg.1 | ENST00000530811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11029060.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11029060.hg.1 | TC11002424.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197786.hg.1 | TC1100012717.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197787.hg.1 | TC1100012717.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000530811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012717.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000227474.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000529801.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000534158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | ENST00000613398.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | NM_001271985.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | NM_031307.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100197789.hg.1 | TC1100012717.hg.1 | Clariom D TC |
