Assay Details
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090738.1 | - | 1 | 1263 | 109 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000267814.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000558789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559230.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000559562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | ENST00000560488.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NM_003104.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007107.hg.1 | NR_034039.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 267302 | - | - |

SORD
Genomic Map