Assay Details
Gene Symbol: MED25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030973.3 | NP_112235.2 | 15 - 16 | 1814 | 163 | |
XM_011527353.1 | XP_011525655.1 | 15 - 16 | 1806 | 163 | ||
GenBank mRNA | AF261072.1 | - | 15 - 16 | 1793 | 163 | |
AF283769.2 | - | 10 - 11 | 1274 | 163 | ||
AF447873.1 | - | 2 - 3 | 2371 | 163 | ||
AJ617479.1 | - | 15 - 16 | 1791 | 163 | ||
AK055844.1 | - | 15 - 16 | 1804 | 100 | ||
AK289460.1 | - | 15 - 16 | 1804 | 163 | ||
AK298897.1 | - | 15 - 16 | 1804 | 100 | ||
AL136746.1 | - | 15 - 16 | 1792 | 163 | ||
AY533507.1 | - | 15 - 16 | 1751 | 163 | ||
BC021543.1 | - | 14 - 15 | 1594 | 163 | ||
BC024312.2 | - | 11 - 12 | 1240 | 163 | ||
BC065297.1 | - | 15 - 16 | 1787 | 6138600 | 163 | |
EU392500.1 | - | 10 - 11 | 1112 | 163 | ||
EU392501.1 | - | 9 - 10 | 956 | 163 | ||
EU392519.1 | - | 8 - 9 | 690 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
