| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000400371.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
ENST00000472805.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009270.hg.1 |
NR_045983.1 |
Refseq |