Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017686.hg.1 |
TC05001265.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017686.hg.1 |
ENST00000397366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017686.hg.1 |
ENST00000397367.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017686.hg.1 |
ENST00000514524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017686.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017686.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010489.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000397366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000397367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000513623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
ENST00000514524.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
NM_001042625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
NM_144647.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172977.hg.1 |
TC0500010489.hg.1 |
Clariom D TC |