Assay Details
Gene Symbol: AMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000036.2 | NP_000027.2 | 8 - 9 | 1242 | 65 | |
NM_001172626.1 | NP_001166097.1 | 7 - 8 | 1230 | 65 | ||
GenBank mRNA | AK097077.1 | - | 7 - 8 | 1230 | 65 | |
AK291349.1 | - | 8 - 9 | 1146 | 65 | ||
AK314252.1 | - | 8 - 9 | 1152 | 65 | ||
M60092.1 | - | 8 - 9 | 1179 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003013.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01046993.hg.1 | TC01003013.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01046993.hg.1 | ENST00000353928.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046993.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046993.hg.1 | ENST00000520113.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046993.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01046993.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100214691.hg.1 | TC0100015350.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000369538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000485564.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | ENST00000520113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_000036.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015350.hg.1 | NM_001172626.1 | Refseq |
