Assay Details
Gene Symbol: C7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138811.3 | NP_620166.3 | 4 - 5 | 945 | 120 | |
XM_005249619.3 | XP_005249676.1 | 4 - 5 | 794 | 120 | ||
XM_005249620.3 | XP_005249677.1 | 5 - 6 | 1325 | 120 | ||
GenBank mRNA | AK097248.1 | - | 5 - 6 | 666 | 120 | |
BC043269.2 | - | 4 - 5 | 1048 | 5297014 | 120 | |
BC060879.1 | - | 4 - 5 | 858 | 5298874 | 120 | |
BI830207.1 | - | 4 - 5 | 482 | 120 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001208.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07018919.hg.1 | TC07001208.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018919.hg.1 | ENST00000283905.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018919.hg.1 | ENST00000409280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018919.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07018918.hg.1 | TC07001208.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07018918.hg.1 | ENST00000283905.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018918.hg.1 | ENST00000409280.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07018918.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174767.hg.1 | TC0700010510.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700174768.hg.1 | TC0700010510.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000283905.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000409280.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000415598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000443822.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | ENST00000444434.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700010510.hg.1 | NM_138811.3 | Refseq |
