| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033740.hg.1 |
TC02004930.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033740.hg.1 |
ENST00000272418.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033740.hg.1 |
NM_031902.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000272418.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000345084.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000461916.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000482568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
NM_031902.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000272418.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000345084.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000461916.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
ENST00000482568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
NM_031902.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200227208.hg.1 |
TC0200016706.hg.1 |
Clariom D TC |