Assay Details
| Gene Symbol: BEND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318539.1 | NP_001305468.1 | 1 - 2 | 488 | 94 | |
| NM_152731.2 | NP_689944.2 | 1 - 2 | 488 | 94 | ||
| XM_011514346.2 | XP_011512648.1 | 1 - 2 | 476 | 94 | ||
| XM_017010408.1 | XP_016865897.1 | 1 - 2 | 476 | 94 | ||
| GenBank mRNA | AK054724.1 | - | 1 - 2 | 173 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000681.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK054724.1,NM_001318539.1,NM_152731.2,XM_011514346.2,XM_017010408.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008125.hg.1 | TC06000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008125.hg.1 | ENST00000322055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008125.hg.1 | ENST00000370748.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008125.hg.1 | ENST00000370750.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008125.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | TC06000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | ENST00000322055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | ENST00000370746.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | ENST00000370748.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | ENST00000370750.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008127.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008124.hg.1 | TC06000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008124.hg.1 | ENST00000322055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008124.hg.1 | ENST00000370750.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008124.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | TC06000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | ENST00000322055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | ENST00000370746.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | ENST00000370748.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | ENST00000370750.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008128.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008126.hg.1 | TC06000681.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008126.hg.1 | ENST00000322055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008126.hg.1 | ENST00000370746.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008126.hg.1 | ENST00000370748.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008126.hg.1 | ENST00000370750.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06008126.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158184.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158184.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158184.hg.1 | TC0600008343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600158185.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158185.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158185.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158185.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158185.hg.1 | TC0600008343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158189.hg.1 | TC0600008343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158186.hg.1 | TC0600008343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158187.hg.1 | TC0600008343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600158190.hg.1 | TC0600008343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370745.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370746.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000370748.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | ENST00000484701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600008343.hg.1 | NM_152731.2 | Refseq |

BEND6
Genomic Map