Assay Details
| Gene Symbol: KCNH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012285.2 | NP_036417.1 | 14 - 15 | 2924 | 103 | |
| XM_017024400.1 | XP_016879889.1 | 14 - 15 | 2920 | 103 | ||
| XM_017024401.1 | XP_016879890.1 | 13 - 14 | 2773 | 103 | ||
| XM_017024402.1 | XP_016879891.1 | 14 - 15 | 2586 | 103 | ||
| GenBank mRNA | AB022698.1 | - | 14 - 15 | 2924 | 103 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001526.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
potassium ion transport
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
phosphorelay sensor kinase activity
voltage-gated potassium channel activity
phosphorelay signal transduction system
potassium ion transport
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
phosphorelay sensor kinase activity
voltage-gated potassium channel activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020354.hg.1 | ENST00000264661.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020354.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020354.hg.1 | TC17001526.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700185064.hg.1 | TC1700010714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000264661.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | ENST00000607371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010714.hg.1 | NM_012285.2 | Refseq |

KCNH4
Genomic Map