Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021974.hg.1 |
TC04001570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021974.hg.1 |
ENST00000280612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021974.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021973.hg.1 |
TC04001570.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021973.hg.1 |
ENST00000280612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021973.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011920.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181832.hg.1 |
TC0400011920.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
ENST00000280612.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
ENST00000509248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
NM_014331.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400181831.hg.1 |
TC0400011920.hg.1 |
Clariom D TC |