Assay Details
| Gene Symbol: CTSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114173.2 | NP_001107645.1 | 1 - 2 | 290 | 68 | |
| NM_001814.5 | NP_001805.3 | 1 - 2 | 290 | 68 | ||
| NM_148170.4 | NP_680475.1 | 1 - 2 | 290 | 68 | ||
| GenBank mRNA | AF234263.1 | - | 1 - 2 | 201 | 68 | |
| AF234264.1 | - | 1 - 2 | 201 | 68 | ||
| AF254757.1 | - | 1 - 2 | 201 | 68 | ||
| AF525032.1 | - | 1 - 2 | 201 | 68 | ||
| AF525033.1 | - | 1 - 2 | 201 | 68 | ||
| AK292117.1 | - | 1 - 2 | 208 | 68 | ||
| AK296190.1 | - | 2 - 3 | 188 | 68 | ||
| AK311923.1 | - | 1 - 2 | 239 | 68 | ||
| BC054028.1 | - | 1 - 2 | 229 | 68 | ||
| BC100891.1 | - | 1 - 2 | 225 | 68 | ||
| BC100892.1 | - | 1 - 2 | 225 | 68 | ||
| BC100893.1 | - | 1 - 2 | 225 | 68 | ||
| BC100894.1 | - | 1 - 2 | 225 | 68 | ||
| BC109386.1 | - | 1 - 2 | 225 | 68 | ||
| BC110071.1 | - | 1 - 2 | 225 | 68 | ||
| BC113850.1 | - | 1 - 2 | 225 | 68 | ||
| BC113897.1 | - | 1 - 2 | 225 | 68 | ||
| BX380095.2 | - | 1 - 2 | 290 | 68 | ||
| BX459226.2 | - | 1 - 2 | 274 | 68 | ||
| BX537913.1 | - | 1 - 2 | 264 | 68 | ||
| CB114425.1 | - | 1 - 2 | 271 | 68 | ||
| X87212.1 | - | 1 - 2 | 208 | 68 | ||
More Information
Set Membership: |
|

CTSC
Genomic Map