Assay Details
| Gene Symbol: CEACAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277163.2 | NP_001264092.1 | 3 - 4 | 630 | 140 | |
| GenBank mRNA | L00693.1 | - | 3 - 4 | 596 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000596544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000596544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | TC1900008161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900162125.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000344550.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000357396.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000415495.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000595255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000596544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000596544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000599662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | ENST00000630848.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001277163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NM_001815.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008161.hg.1 | NR_102333.2 | Refseq |

CEACAM3
Genomic Map