Assay Details
| Gene Symbol: CAMK1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020439.2 | NP_065172.1 | 1 - 2 | 76 | 65 | |
| XM_017001867.1 | XP_016857356.1 | 1 - 2 | 59 | 65 | ||
| GenBank mRNA | AK289675.1 | - | 1 - 2 | 59 | 65 | |
| BC032787.1 | - | 1 - 2 | 43 | 5179957 | 65 | |
| BG715920.1 | - | 1 - 2 | 80 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001748.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK289675.1,BC032787.1,NM_020439.2,XM_017001867.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01027363.hg.1 | TC01001748.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027363.hg.1 | ENST00000009105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027363.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027363.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027361.hg.1 | TC01001748.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027361.hg.1 | ENST00000009105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027361.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027361.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | ENST00000494990.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011451.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184318.hg.1 | TC0100011451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184320.hg.1 | TC0100011451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | ENST00000009105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | ENST00000361322.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | ENST00000423146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | NM_020439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100184321.hg.1 | TC0100011451.hg.1 | Clariom D TC |

CAMK1G
Genomic Map