Assay Details
| Gene Symbol: MTCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018025.3 | NP_001018025.1 | 3 - 4 | 904 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001549.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020786.hg.1 | TC0X001549.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020786.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020786.hg.1 | ENST00000369476.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020786.hg.1 | ENST00000482244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020786.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176851.hg.1 | TC0X00011232.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000362018.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369476.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000369484.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000476116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000482244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | ENST00000504061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018024.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011232.hg.1 | NM_001018025.3 | Refseq |

MTCP1
Genomic Map