Assay Details
Gene Symbol: PABPC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114734.1 | NP_001108206.2 | 2 | 1118 | 157 | |
XM_017007727.1 | XP_016863216.1 | 2 | 1157 | 157 | ||
GenBank mRNA | BX648831.1 | - | 1 | 204 | 157 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001554.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611569 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611516 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450534 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611523 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611531 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611496 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611515 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611524 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450553 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611581 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611514 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450535 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611512 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611509 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611534 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611511 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611510 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450481 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611573 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611501 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450543 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611482 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611532 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611505 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611577 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611568 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611498 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611519 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611502 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611508 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611570 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450561 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611528 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450532 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611526 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611529 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23611565 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03450555 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 116370 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 371600 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 447667 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 309404 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 273690 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P123514 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P123514 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 29407 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33973 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P01083320 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P16867019 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P123514 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P123514 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611579 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611565 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611581 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611564 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611565 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611581 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03450554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611579 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23611536 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P123514 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P16867019 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16867019 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P123514 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 90665 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 83395 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16867019 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16867025 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P906779 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P123514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450506 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450490 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450522 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450538 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450501 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450485 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450554 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450574 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450488 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450474 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450520 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450577 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450508 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15616760 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450524 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450547 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450476 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450530 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450495 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450510 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450544 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450533 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450549 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450498 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450564 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450525 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450552 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450528 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450567 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450536 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450478 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450560 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450504 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450550 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450527 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450511 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450540 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450558 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450519 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450557 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450570 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P03450482 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P16867019 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 238865_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11734404_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11734404_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR04021867.hg.1 | TC04001554.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021867.hg.1 | ENST00000529122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021867.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021866.hg.1 | TC04001554.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021866.hg.1 | ENST00000529122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04021866.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04021865.hg.1 | TC04001554.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04021865.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400181590.hg.1 | ENST00000421491.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400181590.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400181590.hg.1 | ENST00000421491.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400181590.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400181590.hg.1 | TC0400011882.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400181589.hg.1 | ENST00000421491.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400181589.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400181589.hg.1 | ENST00000421491.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400181589.hg.1 | NM_001114734.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400181589.hg.1 | TC0400011882.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 238865_at | - | - |
Human Genome U133 Plus 2.0 Array | 238865_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238865_at5 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024737 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024737 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 110312 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3241262 | NM_001114734.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3241262 | BX648831.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3241262 | NM_001114734.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3241262 | BX648831.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241262 | NM_001114734.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241262 | BX648831.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241262 | NM_001114734.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241262 | BX648831.1 | human_transcriptome |
