Assay Details
Gene Symbol: EMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001423.2 | NP_001414.1 | 3 - 4 | 423 | 75 | |
GenBank mRNA | AK299388.1 | - | 3 - 4 | 305 | 75 | |
AK308505.1 | - | 2 - 3 | 245 | 75 | ||
AK312246.1 | - | 3 - 4 | 297 | 75 | ||
BC047300.1 | - | 3 - 4 | 348 | 4444217 | 75 | |
U43916.1 | - | 2 - 3 | 176 | 75 | ||
U77085.1 | - | 3 - 4 | 392 | 75 | ||
Y07909.1 | - | 3 - 4 | 394 | 75 | ||
Z50751.1 | - | 3 - 4 | 298 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000189.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002387.hg.1 | ENST00000256951.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002387.hg.1 | ENST00000396301.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002387.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002387.hg.1 | ENST00000544053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002387.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002387.hg.1 | TC12000189.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000535134.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000537612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000541935.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000542474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006909.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000256951.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000396301.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000431267.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000536383.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000538364.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000544053.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | ENST00000546357.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | NM_001423.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148333.hg.1 | TC1200006909.hg.1 | Clariom D TC |
