Assay Details
| Gene Symbol: TUBA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032704.4 | NP_116093.1 | 1 - 2 | 183 | 55 | |
| GenBank mRNA | AK223024.1 | - | 1 - 2 | 100 | 55 | |
| BC004949.1 | - | 1 - 2 | 80 | 3617714 | 55 | |
| BC005946.1 | - | 1 - 2 | 100 | 4134187 | 55 | |
| BC011790.1 | - | 1 - 2 | 81 | 4025017 | 55 | |
| BC019298.2 | - | 1 - 2 | 62 | 2905506 | 55 | |
| BC021088.1 | - | 1 - 2 | 61 | 3543481 | 55 | |
| BC033064.1 | - | 1 - 2 | 76 | 3926854 | 55 | |
| BC051297.1 | - | 1 - 2 | 76 | 3832363 | 55 | |
| BC063036.1 | - | 1 - 2 | 122 | 4397636 | 55 | |
| BG469587.1 | - | 1 - 2 | 275 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC004949.1, BC005946.1, BC011790.1, BC019298.2, BC021088.1, BC033064.1, BC051297.1, BC063036.1, NM_032704.4 transcripts.
Set Membership: |
|

TUBA1C
Genomic Map