| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
TC1900011034.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192065.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000263351.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000314121.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000391901.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000487275.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
ENST00000595558.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011034.hg.1 |
NM_001277075.2 |
Refseq |