Assay Details
Gene Symbol: CSRNP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024969.3 | NP_079245.2 | 1 - 2 | 245 | 118 | |
GenBank mRNA | AB063300.1 | - | 1 - 2 | 306 | 118 | |
AK056655.1 | - | 1 - 2 | 242 | 118 | ||
DA500249.1 | - | 1 - 2 | 246 | 118 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000986.hg
Additional Information:
This assay is in the 5'UTR region of the AB063300.1,AK056655.1,NM_024969.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
apoptotic process
negative regulation of phosphatase activity
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription from RNA polymerase II promoter
apoptotic process
negative regulation of phosphatase activity
positive regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | TC02000986.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | ENST00000314499.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | ENST00000342316.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | ENST00000431452.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014458.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02014456.hg.1 | TC02000986.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02014456.hg.1 | ENST00000342316.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02014456.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168890.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168890.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168890.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168890.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168890.hg.1 | TC0200009871.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | ENST00000464503.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009871.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000314499.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000342316.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000409420.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000409664.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | ENST00000421875.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | NM_001172173.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | NM_024969.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200168893.hg.1 | TC0200009871.hg.1 | Clariom D TC |
