Assay Details
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284368.1 | NP_001271297.1 | 1 - 2 | 69 | 58 | |
GenBank mRNA | BM008944.1 | - | 1 - 2 | 57 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001284368.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187885.hg.1 | TC1000012587.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000299166.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370320.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000464651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000466088.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000528425.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000529437.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | ENST00000531418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012587.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | ENST00000370322.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | NM_001284367.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | NM_001284368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | NM_005004.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000187894.hg.1 | TC1000012587.hg.1 | Clariom D TC |
