Assay Details
| Gene Symbol: CLEC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006344.2 | NP_006335.2 | 6 - 7 | 752 | 92 | |
| XM_011523613.1 | XP_011521915.1 | 6 - 7 | 859 | 92 | ||
| XM_011523614.2 | XP_011521916.1 | 7 - 8 | 925 | 92 | ||
| XM_011523615.1 | XP_011521917.1 | 6 - 7 | 859 | 92 | ||
| GenBank mRNA | D50532.1 | - | 6 - 7 | 656 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001074.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014341.hg.1 | ENST00000254868.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014341.hg.1 | ENST00000416562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014341.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014341.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014341.hg.1 | TC17001074.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009612.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000254868.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000416562.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000571664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000576549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | ENST00000576617.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | NM_006344.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | NM_182906.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700174267.hg.1 | TC1700009612.hg.1 | Clariom D TC |

CLEC10A
Genomic Map