Assay Details
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100418.1 | NP_001093888.1 | 2 - 3 | 293 | 131 | |
GenBank mRNA | AK000857.1 | - | 2 - 3 | 180 | 131 | |
BC012078.1 | - | 2 - 3 | 182 | 4661993 | 131 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000361.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19004781.hg.1 | ENST00000358607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19004781.hg.1 | ENST00000450195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19004781.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004781.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004781.hg.1 | TC19000361.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004780.hg.1 | ENST00000358607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19004780.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004780.hg.1 | TC19000361.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | TC1900007398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154365.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | TC1900007398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154364.hg.1 | NM_001100418.1 | Refseq |
