Assay Details
Gene Symbol: C19orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100418.1 | NP_001093888.1 | 3 - 4 | 571 | 135 | |
NM_001100419.1 | NP_001093889.1 | 3 - 4 | 505 | 135 | ||
XM_005259977.1 | XP_005260034.1 | 1 - 2 | 492 | 135 | ||
GenBank mRNA | AK000857.1 | - | 3 - 4 | 459 | 135 | |
AK054670.1 | - | 3 - 4 | 490 | 135 | ||
AK091925.1 | - | 2 - 3 | 793 | 135 | ||
BC012078.1 | - | 3 - 4 | 460 | 4661993 | 135 | |
BI836487.1 | - | 3 - 4 | 382 | 135 | ||
KP143545.1 | - | 3 - 4 | 365 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000361.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004782.hg.1 | ENST00000358607.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19004782.hg.1 | ENST00000450195.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19004782.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004782.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19004782.hg.1 | TC19000361.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | TC1900007398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154366.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | TC1900007398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900154369.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000358607.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000450195.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595077.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000595490.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000597371.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000598375.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000600997.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | ENST00000601736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100418.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007398.hg.1 | NM_001100419.1 | Refseq |
