Assay Details
| Gene Symbol: POLR1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282766.1 | NP_001269695.1 | 3 - 4 | 373 | 63 | |
| NM_022490.2 | NP_071935.1 | 3 - 4 | 373 | 63 | ||
| XM_005251547.2 | XP_005251604.1 | 2 - 3 | 618 | 63 | ||
| GenBank mRNA | AK023452.1 | - | 3 - 4 | 360 | 63 | |
| AK024032.1 | - | 3 - 4 | 349 | 63 | ||
| AK091294.1 | - | 2 - 3 | 734 | 63 | ||
| AK225050.1 | - | 3 - 4 | 346 | 63 | ||
| AK301347.1 | - | 3 - 4 | 363 | 63 | ||
| AK303167.1 | - | 3 - 4 | 360 | 63 | ||
| BC001337.1 | - | 3 - 4 | 340 | 3461710 | 63 | |
| BC014331.1 | - | 3 - 4 | 374 | 4042929 | 63 | |
| CR457313.1 | - | 3 - 4 | 260 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09002930.hg
Additional Information:
This assay is in the 5'UTR region of the AK303167.1,NM_001282766.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
RNA polymerase I transcriptional preinitiation complex assembly at the promoter for the nuclear large rRNA transcript
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
rRNA transcription
positive regulation of gene expression, epigenetic
RNA polymerase I activity
RNA polymerase I transcription factor binding
DNA binding
RNA polymerase I transcriptional preinitiation complex assembly at the promoter for the nuclear large rRNA transcript
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
rRNA transcription
positive regulation of gene expression, epigenetic
RNA polymerase I activity
RNA polymerase I transcription factor binding
DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09002508.hg.1 | TC09002930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002508.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002508.hg.1 | ENST00000377798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002508.hg.1 | ENST00000421959.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002508.hg.1 | ENST00000442009.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002508.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002511.hg.1 | TC09002930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002511.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002511.hg.1 | ENST00000377798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002511.hg.1 | ENST00000442009.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002511.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002510.hg.1 | TC09002930.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002510.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002510.hg.1 | ENST00000377798.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002510.hg.1 | ENST00000421959.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002510.hg.1 | ENST00000442009.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002510.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183885.hg.1 | TC0900012124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900183886.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183886.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183886.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183886.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183886.hg.1 | TC0900012124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | ENST00000377792.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | ENST00000377798.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | ENST00000442009.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | NM_001282766.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | NM_022490.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900183888.hg.1 | TC0900012124.hg.1 | Clariom D TC |

POLR1E
Genomic Map