Assay Details
Gene Symbol: POLR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282766.1 | NP_001269695.1 | 8 - 9 | 848 | 103 | |
NM_022490.2 | NP_071935.1 | 7 - 8 | 754 | 103 | ||
XM_005251547.2 | XP_005251604.1 | 6 - 7 | 999 | 103 | ||
GenBank mRNA | AK023452.1 | - | 7 - 8 | 741 | 103 | |
AK024032.1 | - | 7 - 8 | 730 | 103 | ||
AK091294.1 | - | 6 - 7 | 1115 | 103 | ||
AK225050.1 | - | 7 - 8 | 727 | 103 | ||
AK302345.1 | - | 6 - 7 | 731 | 103 | ||
AK303167.1 | - | 8 - 9 | 835 | 103 | ||
BC001337.1 | - | 7 - 8 | 721 | 3461710 | 103 | |
BC014331.1 | - | 7 - 8 | 755 | 4042929 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09002930.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
Function(s)
Process(es)
RNA polymerase I transcriptional preinitiation complex assembly at the promoter for the nuclear large rRNA transcript
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
rRNA transcription
positive regulation of gene expression, epigenetic
RNA polymerase I activity
RNA polymerase I transcription factor binding
DNA binding
RNA polymerase I transcriptional preinitiation complex assembly at the promoter for the nuclear large rRNA transcript
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
rRNA transcription
positive regulation of gene expression, epigenetic
RNA polymerase I activity
RNA polymerase I transcription factor binding
DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09002515.hg.1 | TC09002930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002515.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002515.hg.1 | ENST00000377798.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002515.hg.1 | ENST00000442009.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002515.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900183891.hg.1 | TC0900012124.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000377798.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | ENST00000442009.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_001282766.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012124.hg.1 | NM_022490.2 | Refseq |
