| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009688.hg.1 |
ENST00000301019.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009688.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009688.hg.1 |
TC16000686.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165910.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165910.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165910.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165910.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165910.hg.1 |
TC1600008845.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000562747.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600008845.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
ENST00000301019.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
ENST00000569140.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
NM_030928.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600165911.hg.1 |
TC1600008845.hg.1 |
Clariom D TC |