Assay Details
Gene Symbol: KCTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319294.1 | NP_001306223.1 | 14 - 15 | 1766 | 72 | |
NM_001319295.1 | NP_001306224.1 | 14 - 15 | 1838 | 72 | ||
NM_016121.4 | NP_057205.2 | 14 - 15 | 1766 | 72 | ||
XM_005273158.1 | XP_005273215.1 | 14 - 15 | 2032 | 72 | ||
GenBank mRNA | AK296275.1 | - | 12 - 13 | 1299 | 72 | |
AK316272.1 | - | 10 - 11 | 1167 | 72 | ||
AL834389.1 | - | 8 - 9 | 990 | 72 | ||
BC013868.2 | - | 14 - 15 | 1753 | 72 | ||
BC094879.1 | - | 14 - 15 | 1715 | 3078971 | 72 | |
BC117188.1 | - | 14 - 15 | 1648 | 40125739 | 72 | |
BC126173.1 | - | 14 - 15 | 1648 | 8991889 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001793.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01028084.hg.1 | TC01001793.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01028084.hg.1 | ENST00000259154.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028084.hg.1 | ENST00000366946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028084.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185390.hg.1 | TC0100011592.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
