Assay Details
Gene Symbol: KCTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319294.1 | NP_001306223.1 | 1 - 2 | 391 | 101 | |
NM_001319295.1 | NP_001306224.1 | 1 - 2 | 391 | 101 | ||
NM_016121.4 | NP_057205.2 | 1 - 2 | 391 | 101 | ||
XM_005273158.1 | XP_005273215.1 | 1 - 2 | 585 | 101 | ||
GenBank mRNA | BC013868.2 | - | 1 - 2 | 378 | 101 | |
BC094879.1 | - | 1 - 2 | 340 | 3078971 | 101 | |
BC117188.1 | - | 1 - 2 | 273 | 40125739 | 101 | |
BC126173.1 | - | 1 - 2 | 273 | 8991889 | 101 | |
HY029594.1 | - | 1 - 2 | 392 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001793.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001319295.1,XM_005273158.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01028063.hg.1 | TC01001793.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01028063.hg.1 | ENST00000259154.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028063.hg.1 | ENST00000366945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028063.hg.1 | ENST00000366946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028063.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01028065.hg.1 | TC01001793.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01028065.hg.1 | ENST00000259154.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028065.hg.1 | ENST00000366945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028065.hg.1 | ENST00000366946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028065.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01028066.hg.1 | TC01001793.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01028066.hg.1 | ENST00000259154.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028066.hg.1 | ENST00000366945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028066.hg.1 | ENST00000366946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028066.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01028064.hg.1 | TC01001793.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01028064.hg.1 | ENST00000259154.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028064.hg.1 | ENST00000366945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028064.hg.1 | ENST00000366946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028064.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185367.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185367.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185367.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185367.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185367.hg.1 | TC0100011592.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100185366.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185366.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185366.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185366.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185366.hg.1 | TC0100011592.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185369.hg.1 | TC0100011592.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185368.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185368.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185368.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185368.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185368.hg.1 | TC0100011592.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100185365.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185365.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185365.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185365.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185365.hg.1 | TC0100011592.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 168268 | - | - |
