Assay Details
Gene Symbol: KCTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319294.1 | NP_001306223.1 | 3 - 4 | 488 | 75 | |
NM_001319295.1 | NP_001306224.1 | 3 - 4 | 488 | 75 | ||
NM_016121.4 | NP_057205.2 | 3 - 4 | 488 | 75 | ||
XM_005273158.1 | XP_005273215.1 | 3 - 4 | 682 | 75 | ||
GenBank mRNA | BC013868.2 | - | 3 - 4 | 475 | 75 | |
BC094879.1 | - | 3 - 4 | 437 | 3078971 | 75 | |
BC117188.1 | - | 3 - 4 | 370 | 40125739 | 75 | |
BC126173.1 | - | 3 - 4 | 370 | 8991889 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001793.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001319295.1,XM_005273158.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01028068.hg.1 | TC01001793.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01028068.hg.1 | ENST00000259154.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028068.hg.1 | ENST00000366945.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028068.hg.1 | ENST00000366946.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01028068.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000452413.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000465650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | ENST00000495537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011592.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | ENST00000259154.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | ENST00000448333.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | NM_016121.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100185371.hg.1 | TC0100011592.hg.1 | Clariom D TC |
