Assay Details
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052928.2 | NP_443160.2 | 8 - 9 | 2190 | 56 | |
XM_017024121.1 | XP_016879610.1 | 9 - 10 | 2435 | 56 | ||
GenBank mRNA | AB067523.1 | - | 4 - 5 | 1434 | 56 | |
AK057769.1 | - | 4 - 5 | 622 | 56 | ||
AK092149.1 | - | 4 - 5 | 1095 | 56 | ||
AK095369.1 | - | 8 - 9 | 2190 | 56 | ||
BC035077.2 | - | 8 - 9 | 2109 | 5260399 | 56 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000984.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17013176.hg.1 | ENST00000305513.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013176.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013176.hg.1 | TC17000984.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17013177.hg.1 | ENST00000305513.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17013177.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17013177.hg.1 | TC17000984.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000570368.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000571854.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000573937.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000576633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | ENST00000582190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009402.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172024.hg.1 | TC1700009402.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | ENST00000305513.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | ENST00000476292.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | ENST00000491788.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | NM_052928.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700172023.hg.1 | TC1700009402.hg.1 | Clariom D TC |
