Assay Details
| Gene Symbol: MIOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017584.5 | NP_060054.4 | 2 - 3 | 211 | 84 | |
| XM_011530705.1 | XP_011529007.1 | 3 - 4 | 2561 | 84 | ||
| GenBank mRNA | AF197129.1 | - | 1 - 2 | 97 | 84 | |
| AY064416.1 | - | 2 - 3 | 124 | 84 | ||
| AY738258.1 | - | 1 - 2 | 97 | 84 | ||
| BC012025.1 | - | 2 - 3 | 211 | 4454512 | 84 | |
| BC064395.1 | - | 2 - 3 | 119 | 84 | ||
| BC073848.1 | - | 2 - 3 | 131 | 5186354 | 84 | |
| CR456478.1 | - | 2 - 3 | 156 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000427.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22008089.hg.1 | ENST00000216075.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008089.hg.1 | ENST00000395732.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008089.hg.1 | ENST00000395733.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008089.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22008089.hg.1 | TC22000427.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | TC2200007809.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155461.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000216075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395732.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000395733.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | ENST00000451761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007809.hg.1 | NM_017584.5 | Refseq |

MIOX
Genomic Map