Assay Details
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011523910.2 | XP_011522212.1 | 1 - 2 | 405 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011523910.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
