Assay Details
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005256667.4 | XP_005256724.1 | 2 - 3 | 245 | 96 | |
XM_011523910.2 | XP_011522212.1 | 2 - 3 | 506 | 96 | ||
XM_011523911.2 | XP_011522213.1 | 2 - 3 | 346 | 96 | ||
XM_017024709.1 | XP_016880198.1 | 3 - 4 | 598 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005256667.4, XM_011523910.2, XM_011523911.2, XM_017024709.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000319004.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000437269.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000570364.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000573482.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000574958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576383.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | ENST00000576778.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009327.hg.1 | NM_015721.2 | Refseq |
