Assay Details
| Gene Symbol: TYMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB077207.1 | - | 1 - 2 | 204 | 81 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000584122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | ENST00000584122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323224.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323250.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000323274.14 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000579128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000581920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000584122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | ENST00000584122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006448.hg.1 | NM_001071.2 | Refseq |

TYMS
Genomic Map