Assay Details
Gene Symbol: SNRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017719.4 | NP_060189.3 | 2 - 3 | 227 | 184 | |
GenBank mRNA | BP281284.1 | - | 2 - 3 | 199 | 184 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_017719.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
