Assay Details
| Gene Symbol: USE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018467.3 | NP_060937.1 | 2 - 3 | 213 | 82 | |
| XM_017026975.1 | XP_016882464.1 | 3 - 4 | 923 | 82 | ||
| GenBank mRNA | AB097050.1 | - | 2 - 3 | 184 | 82 | |
| AF220052.1 | - | 2 - 3 | 184 | 82 | ||
| AK074683.1 | - | 2 - 3 | 169 | 82 | ||
| BC006005.1 | - | 2 - 3 | 169 | 4279653 | 82 | |
| BC008455.1 | - | 2 - 3 | 161 | 4247241 | 82 | |
| BI758045.1 | - | 2 - 3 | 200 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000328.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004272.hg.1 | ENST00000263897.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004272.hg.1 | ENST00000379776.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004272.hg.1 | ENST00000445667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004272.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004272.hg.1 | TC19000328.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | TC1900007313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153041.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000263897.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000445667.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000593597.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594399.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000594724.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000595101.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000596136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000600970.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601592.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | ENST00000601662.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | NM_018467.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007313.hg.1 | NM_018467.3 | Refseq |

USE1
Genomic Map