Assay Details
| Gene Symbol: LMNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032737.3 | NP_116126.3 | 8 - 9 | 1515 | 71 | |
| GenBank mRNA | BC006551.2 | - | 8 - 9 | 1480 | 2961134 | 71 |
| BT007441.1 | - | 8 - 9 | 1420 | 71 | ||
| KP137589.1 | - | 5 - 6 | 1069 | 71 | ||
| M94362.1 | - | 7 - 8 | 1165 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001030.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19014069.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014069.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014069.hg.1 | TC19001030.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014070.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014070.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19014070.hg.1 | TC19001030.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000475819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000475819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000534495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000534495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000534495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000614319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | ENST00000614319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NR_106958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011841.hg.1 | NR_106958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203529.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203529.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203529.hg.1 | TC1900011841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203529.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203529.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000475819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000475819.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000490554.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000527409.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000534495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000534495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000534495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000614319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | ENST00000614319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NR_106958.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011841.hg.1 | NR_106958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203528.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203528.hg.1 | NM_032737.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900203528.hg.1 | TC1900011841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900203528.hg.1 | ENST00000325327.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900203528.hg.1 | NM_032737.3 | Refseq |

LMNB2
Genomic Map