Assay Details
| Gene Symbol: PRPF40A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017892.3 | NP_060362.3 | 24 - 25 | 2589 | 144 | |
| XM_005246676.4 | XP_005246733.3 | 24 - 25 | 3109 | 144 | ||
| XM_005246677.4 | XP_005246734.2 | 24 - 25 | 2597 | 144 | ||
| XM_011511446.2 | XP_011509748.1 | 23 - 24 | 3067 | 144 | ||
| XM_011511447.2 | XP_011509749.1 | 24 - 25 | 3121 | 144 | ||
| XM_011511448.2 | XP_011509750.1 | 23 - 24 | 2543 | 144 | ||
| XM_011511449.2 | XP_011509751.1 | 26 - 27 | 2951 | 144 | ||
| XM_017004453.1 | XP_016859942.1 | 24 - 25 | 3121 | 144 | ||
| XM_017004454.1 | XP_016859943.1 | 23 - 24 | 3055 | 144 | ||
| XM_017004455.1 | XP_016859944.1 | 23 - 24 | 2531 | 144 | ||
| XM_017004456.1 | XP_016859945.1 | 25 - 26 | 2787 | 144 | ||
| XM_017004457.1 | XP_016859946.1 | 26 - 27 | 2938 | 144 | ||
| XM_017004458.1 | XP_016859947.1 | 24 - 25 | 2697 | 144 | ||
| XM_017004459.1 | XP_016859948.1 | 24 - 25 | 2806 | 144 | ||
| XM_017004460.1 | XP_016859949.1 | 24 - 25 | 2722 | 144 | ||
| XM_017004461.1 | XP_016859950.1 | 24 - 25 | 2794 | 144 | ||
| GenBank mRNA | AF151059.1 | - | 8 - 9 | 721 | 144 | |
| AF155096.1 | - | 10 - 11 | 1026 | 144 | ||
| AK000592.1 | - | 6 - 7 | 521 | 144 | ||
| AK315187.1 | - | 6 - 7 | 553 | 144 | ||
| BC027178.1 | - | 5 - 6 | 423 | 144 | ||
More Information
Set Membership: |
|

PRPF40A
Genomic Map