Assay Details
| Gene Symbol: SGSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301849.1 | NP_001288778.1 | 2 - 3 | 242 | 87 | |
| XM_005261575.1 | XP_005261632.1 | 2 - 3 | 220 | 87 | ||
| XM_011530160.1 | XP_011528462.1 | 2 - 3 | 219 | 87 | ||
| XM_017028775.1 | XP_016884264.1 | 2 - 3 | 206 | 87 | ||
| XM_017028776.1 | XP_016884265.1 | 2 - 3 | 213 | 87 | ||
| XM_017028777.1 | XP_016884266.1 | 2 - 3 | 213 | 87 | ||
| XM_017028778.1 | XP_016884267.1 | 2 - 3 | 220 | 87 | ||
| XM_017028779.1 | XP_016884268.1 | 2 - 3 | 219 | 87 | ||
| XM_017028780.1 | XP_016884269.1 | 2 - 3 | 208 | 87 | ||
| GenBank mRNA | AK297599.1 | - | 2 - 3 | 173 | 87 | |
| AK301042.1 | - | 2 - 3 | 155 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK297599.1, AK301042.1, NM_001301849.1, XM_005261575.1, XM_011530160.1, XM_017028775.1, XM_017028776.1, XM_017028777.1, XM_017028778.1, XM_017028779.1, XM_017028780.1 transcripts.
Set Membership: |
|

SGSM3
Genomic Map