Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004656.hg.1 |
ENST00000266085.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004656.hg.1 |
ENST00000382049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004656.hg.1 |
ENST00000538671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004656.hg.1 |
NM_000362.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22004656.hg.1 |
TC22000248.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200150352.hg.1 |
TC2200007150.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200150352.hg.1 |
ENST00000266085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150352.hg.1 |
NM_000362.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200150352.hg.1 |
ENST00000266085.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200150352.hg.1 |
NM_000362.4 |
Refseq |