Assay Details
Gene Symbol: SEPT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF285108.1 | - | 2 - 3 | 219 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396417.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396425.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000396426.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000406029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000449288.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | ENST00000460267.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_019106.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007505.hg.1 | NM_145733.2 | Refseq |
