| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022938.hg.1 |
ENST00000377264.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022938.hg.1 |
ENST00000421419.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022938.hg.1 |
NM_015104.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11022938.hg.1 |
TC11001918.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
ENST00000377264.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
ENST00000418259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
NM_015104.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
ENST00000377264.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
ENST00000418259.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
NM_015104.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100185634.hg.1 |
TC1100011192.hg.1 |
Clariom D TC |