Assay Details
| Gene Symbol: DLEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC033120.1 | - | 1 | 748 | 118 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | potassium channel regulator | KCNRG | DLTET | -18.7kb | Anti-sense | Chr. 13 - 50015254 - 50020922 on NCBI Build GRCh38 |
| Downstream | microRNA 16-1 | MIR16-1 |
MIRN16-1, miRNA16-1, mir-16-1 |
7.9kb | Sense | Chr. 13 - 50048973 - 50049061 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DLEU2
Genomic Map