| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035000.hg.1 |
TC03001956.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035000.hg.1 |
ENST00000334256.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03035000.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196969.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300196969.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196969.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300196969.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300196969.hg.1 |
TC0300012970.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000334256.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000469804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
ENST00000483437.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300012970.hg.1 |
NM_002268.4 |
Refseq |