Assay Details
| Gene Symbol: PIK3CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006219.2 | NP_006210.1 | 5 - 6 | 991 | 126 | |
| XM_005247530.2 | XP_005247587.1 | 7 - 8 | 1340 | 126 | ||
| XM_006713659.3 | XP_006713722.1 | 6 - 7 | 1211 | 126 | ||
| XM_011512895.2 | XP_011511197.1 | 8 - 9 | 1336 | 126 | ||
| XM_011512896.1 | XP_011511198.1 | 4 - 5 | 656 | 126 | ||
| XM_017006619.1 | XP_016862108.1 | 6 - 7 | 1014 | 126 | ||
| GenBank mRNA | BC114432.1 | - | 5 - 6 | 979 | 40008544 | 126 |
| S67334.1 | - | 5 - 6 | 975 | 126 | ||
More Information
Updated Mapping Information
Set Membership: |
|

PIK3CB
Genomic Map